missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-[3,5-di(trifluoromethyl)phenyl]ethan-1-one, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Linear Formula | (CF3)2C6H3C(O)CH2Br |
Quantity | 25 g |
Formula Weight | 335.04 |
Percent Purity | 95% |
Chemical Name or Material | 2-Bromo-1-[3, 5-di(trifluoromethyl)phenyl]ethan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'