missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(2-thienyl)-1-ethanone, Technical Grade, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 205.07 |
Percent Purity | 90% |
Chemical Name or Material | 2-Bromo-1-(2-thienyl)-1-ethanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'