missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-one, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 95% |
Quantity | 250 mg |
IUPAC Name | 2-bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-one |
Formula Weight | 257.08 |
Percent Purity | 95% |
Chemical Name or Material | 2-Bromo-1-(2, 3-dihydro-1, 4-benzodioxin-6-yl)ethan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'