missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Boc-aminomethyl)-4-fluorobenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62345.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15417316

  • £333.10 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(Boc-aminomethyl)-4-fluorobenzeneboronic acid pinacol ester
351.22
2-(Boc-aminomethyl)-4-fluorophenylboronic acid pinacol ester

Specifications

2-(Boc-aminomethyl)-4-fluorobenzeneboronic acid pinacol ester
96%
1g
Solid
351.22
2-(Boc-aminomethyl)-4-fluorophenylboronic acid pinacol ester
Videos
SDS