missing translation for 'onlineSavingsMsg'
Learn More

2-(BOC-Amino)ethanethiol, 97%, Thermo Scientific Chemicals

Catalog Number 10668224
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
67385-09-5
C7H15NO2S
177.26
MFCD00274335
GSJJCZSHYJNRPN-UHFFFAOYSA-N
2-boc-amino ethanethiol, boc-cysteamine, tert-butyl n-2-mercaptoethyl carbamate, tert-butyl n-2-sulfanylethyl carbamate, 2-bocamino ethanethiol, tert-butyl 2-mercaptoethyl carbamate, carbamic acid, 2-mercaptoethyl-, 1,1-dimethylethyl ester, n-boc-aet, tert-butoxy-n-2-sulfanylethyl carboxamide, n-boc-cysteamine
3017761
tert-butyl N-(2-sulfanylethyl)carbamate
CC(C)(C)OC(=O)NCCS
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
67385-09-5
C7H15NO2S
177.26
MFCD00274335
GSJJCZSHYJNRPN-UHFFFAOYSA-N
2-boc-amino ethanethiol, boc-cysteamine, tert-butyl n-2-mercaptoethyl carbamate, tert-butyl n-2-sulfanylethyl carbamate, 2-bocamino ethanethiol, tert-butyl 2-mercaptoethyl carbamate, carbamic acid, 2-mercaptoethyl-, 1,1-dimethylethyl ester, n-boc-aet, tert-butoxy-n-2-sulfanylethyl carboxamide, n-boc-cysteamine
3017761
tert-butyl N-(2-sulfanylethyl)carbamate
CC(C)(C)OC(=O)NCCS

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 68.0°C (0.3 mmHg)
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Assay Percent Range 96% min. (GC)
Infrared Spectrum Authentic
Linear Formula (CH3)3COC(O)NH(CH2)2SH
Packaging Glass Bottle
Refractive Index 1.4730 to 1.4770
Formula Weight 177.27
Percent Purity 97%
Density 1.0490g/mL
Quantity 5 mL
Flash Point 109°C
Specific Gravity 1.049
Physical Form Low Melting Solid or Viscous Liquid
Chemical Name or Material 2-(BOC-Amino)ethanethiol
Show More Show Less
compliance-icons
Product Identifier
  • 2-(BOC-Amino)ethanethiol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'