missing translation for 'onlineSavingsMsg'
Learn More

2-(Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino)benzaldehyde, 97%, ACROS Organics™

£50.18 - £63.40

Chemical Identifiers

CAS 1202865-21-1
Molecular Formula C37H51O3P
Molecular Weight (g/mol) 574.78
MDL Number MFCD15143663
InChI Key PAWZHFURVJBYGV-UHFFFAOYSA-N
Synonym 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl benzaldehyde, acmc-20ao6l, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, min
PubChem CID 71310909
IUPAC Name 2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanylbenzaldehyde
SMILES CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC=CC=C2C=O)C3=CC(=C(C(=C3)C(C)(C)C)OC)C(C)(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10037999 SDS View Documents Acros Organics
435132500
250mg £50.18
250mg View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
10371100 SDS View Documents Acros Organics
435131000
100mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Specifications

Chemical Identifiers

1202865-21-1
574.78
PAWZHFURVJBYGV-UHFFFAOYSA-N
71310909
CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC=CC=C2C=O)C3=CC(=C(C(=C3)C(C)(C)C)OC)C(C)(C)C
C37H51O3P
MFCD15143663
2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl benzaldehyde, acmc-20ao6l, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, min
2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanylbenzaldehyde

Specifications

2-(Bis(3, 5-di-tert-Butyl-4-methoxyphenyl)phosphino)benzaldehyde
Authentic
Yellow
250mg
96.0
96% min. (GC)
MFCD15143663
PAWZHFURVJBYGV-UHFFFAOYSA-N
2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanylbenzaldehyde
71310909
Powder
74.22 to 80.41% C
(1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure
123.0°C to 124.5°C
1202865-21-1
100.0
C37H51O3P
2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl benzaldehyde, acmc-20ao6l, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, min
CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC=CC=C2C=O)C3=CC(=C(C(=C3)C(C)(C)C)OC)C(C)(C)C
574.78
574.78
97%
Safety and Handling

Safety and Handling

Product Identifier
  • 2-(Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino)benzaldehyde

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.