missing translation for 'onlineSavingsMsg'
Learn More

2-Biphenylcarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-66945
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
947-84-2
C13H10O2
198.22
MFCD00002463
ILYSAKHOYBPSPC-UHFFFAOYSA-N
2-biphenylcarboxylic acid, 1,1'-biphenyl-2-carboxylic acid, o-phenylbenzoic acid, biphenyl-2-carboxylic acid, biphenylcarboxylic acid, 1,1'-biphenyl carboxylic acid, diphenyl-2-carboxylic acid, phenylbenzoic acid
70357
2-phenylbenzoic acid
C1=CC=C(C=C1)C2=CC=CC=C2C(=O)O
This item is not returnable. View return policy
947-84-2
C13H10O2
198.22
MFCD00002463
ILYSAKHOYBPSPC-UHFFFAOYSA-N
2-biphenylcarboxylic acid, 1,1'-biphenyl-2-carboxylic acid, o-phenylbenzoic acid, biphenyl-2-carboxylic acid, biphenylcarboxylic acid, 1,1'-biphenyl carboxylic acid, diphenyl-2-carboxylic acid, phenylbenzoic acid
70357
2-phenylbenzoic acid
C1=CC=C(C=C1)C2=CC=CC=C2C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 110.0°C to 114.0°C
Color White to Beige
Boiling Point 199.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5C6H4CO2H
Quantity 25 g
Beilstein 09,669
Solubility Information Solubility in water: insoluble.
Formula Weight 198.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Biphenylcarboxylic acid, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'