missing translation for 'onlineSavingsMsg'
Learn More

2-Bibenzylcarboxylic acid, 98%, ACROS Organics™

Brand:  Acros Organics 348810250

Code : 29

Additional Details : CAS Number : 4890-85-1 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10748542

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Bibenzylcarboxylic acid
97.5
≥97.5%
C15H14O2
226.27
2-Phenethylbenzoic acid
4890-85-1
100.0
98%
C6H5(CH2)2C6H4CO2H
MFCD00002485

Specifications

2-Bibenzylcarboxylic acid
97.5
Authentic
127° to 132°C
Crystalline Powder
98%
C15H14O2
226.27
2-Phenethylbenzoic acid
4890-85-1
100.0
Glass bottle
White to off-white
≥97.5%
25g
C6H5(CH2)2C6H4CO2H
MFCD00002485
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bibenzylcarboxylic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS