missing translation for 'onlineSavingsMsg'
Learn More

2-BFI hydrochloride, Tocris Bioscience™

Potent, selective I2 ligand. Putative agonist

Brand:  Tocris (Bio-Techne) 0348/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13467573

  • £151.41 / 10mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

High affinity I2 ligand (Ki = 9.8 nM). Putative I2 agonist; potentiates morphine antinociception. Produces ipsiversive rotational behavior in rats with a full 6-OHDA lesion of the nigrostriatal tract.
Specifications

Chemical Identifiers

89196-95-2
222.672
2-2-benzofuranyl-2-imidazoline hydrochloride, 2-bfi hydrochloride, 2-benzofuran-2-yl-2-imidazoline hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole hydrochloride, 2-bfi hydrochloride hplc, 2-2-benzo b furanyl-2-imidazoline hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole,hydrochloride, 2-benzofuran-2-yl-4,5-dihydro-1h-imidazole hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole-hydrogen chloride 1/1
2-(1-benzofuran-2-yl)-4,5-dihydro-1H-imidazole;hydrochloride
C11H11ClN2O
RFNFFVDVGWOSNZ-UHFFFAOYSA-N
45073465
C1CN=C(N1)C2=CC3=CC=CC=C3O2.Cl

Specifications

10mg
89196-95-2
2-2-benzofuranyl-2-imidazoline hydrochloride, 2-bfi hydrochloride, 2-benzofuran-2-yl-2-imidazoline hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole hydrochloride, 2-bfi hydrochloride hplc, 2-2-benzo b furanyl-2-imidazoline hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole,hydrochloride, 2-benzofuran-2-yl-4,5-dihydro-1h-imidazole hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole-hydrogen chloride 1/1
C1CN=C(N1)C2=CC3=CC=CC=C3O2.Cl
222.672
222.67
2-BFI hydrochloride
C11H11ClN2O
RFNFFVDVGWOSNZ-UHFFFAOYSA-N
2-(1-benzofuran-2-yl)-4,5-dihydro-1H-imidazole;hydrochloride
45073465
>98%
Videos
Documents