missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Benzylthiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62209.06

Code : NEW

Additional Details : CAS Number : 50739-30-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15488748

  • £321.80 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Benzylthiazolidine-4-carboxylic acid
50739-30-5
223.29
MFCD09936775

Specifications

2-Benzylthiazolidine-4-carboxylic acid
Solid
223.29
5g
50739-30-5
97%
MFCD09936775
Videos
SDS