missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxyethanol, 98%, Thermo Scientific Chemicals

Catalog Number 10509653
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
622-08-2
C9H12O2
152.19
MFCD00002868
CUZKCNWZBXLAJX-UHFFFAOYSA-N
2-benzyloxy ethanol, 2-benzyloxyethanol, ethanol, 2-phenylmethoxy, ethylene glycol monobenzyl ether, 2-benzyloxy-1-ethanol, benzyl cellosolve, glycol benzyl ether, ethanol, 2-benzyloxy, glycol monobenzyl ether
12141
2-phenylmethoxyethanol
OCCOCC1=CC=CC=C1
This item is not returnable. View return policy
622-08-2
C9H12O2
152.19
MFCD00002868
CUZKCNWZBXLAJX-UHFFFAOYSA-N
2-benzyloxy ethanol, 2-benzyloxyethanol, ethanol, 2-phenylmethoxy, ethylene glycol monobenzyl ether, 2-benzyloxy-1-ethanol, benzyl cellosolve, glycol benzyl ether, ethanol, 2-benzyloxy, glycol monobenzyl ether
12141
2-phenylmethoxyethanol
OCCOCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.0700g/mL
Boiling Point 265°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OCH2CH2OH
Refractive Index 1.5200 to 1.522
Quantity 1 g
Specific Gravity 1.07
Formula Weight 152.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Benzyloxyethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'