missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzyloxy)phenol, 97%, Thermo Scientific Chemicals

Catalog Number p-4258763
Click to view available options
Quantity:
5 g
10 g
This item is not returnable. View return policy
6272-38-4
C13H12O2
200.237
CCZCXFHJMKINPE-UHFFFAOYSA-N
2-benzyloxy phenol, 2-benzyloxyphenol, phenol, 2-phenylmethoxy, benzyloxyphenol, benzyl o-hydroxyphenyl ether, 2-phenylmethoxy phenol, 2-benzyloxy-phenol, o-benzyloxy phenol
80459
2-phenylmethoxyphenol
C1=CC=C(C=C1)COC2=CC=CC=C2O
This item is not returnable. View return policy
6272-38-4
C13H12O2
200.237
CCZCXFHJMKINPE-UHFFFAOYSA-N
2-benzyloxy phenol, 2-benzyloxyphenol, phenol, 2-phenylmethoxy, benzyloxyphenol, benzyl o-hydroxyphenyl ether, 2-phenylmethoxy phenol, 2-benzyloxy-phenol, o-benzyloxy phenol
80459
2-phenylmethoxyphenol
C1=CC=C(C=C1)COC2=CC=CC=C2O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 35.0°C to 40.0°C
Color Colorless to Yellow
Density 1.1400g/mL
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula C6H5CH2OC6H4OH
Refractive Index 1.5906
Quantity 5 g
Specific Gravity 1.14
Formula Weight 200.24
Percent Purity 97%
Physical Form Liquid or Low Melting Mass
Chemical Name or Material 2-(Benzyloxy)phenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Benzyloxyphenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'