missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-{[(Benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid, 97%, Maybridge™
Click to view available options
Quantity:
10 g
25 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | C6H5CH2OC(O)NHCH(CO2H)CH2C6H4OH |
Quantity | 10 g |
Formula Weight | 315.32 |
Percent Purity | 97% |
Chemical Name or Material | 2-{[(Benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'