missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Benzyloxy-5-fluorobenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62613.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447636

  • £302.30 / 1g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Benzyloxy-5-fluorobenzeneboronic acid pinacol ester
328.19
2-Benzyloxy-5-fluorophenylboronic acid pinacol ester

Specifications

2-Benzyloxy-5-fluorobenzeneboronic acid pinacol ester
Solid
328.19
2-Benzyloxy-5-fluorophenylboronic acid pinacol ester
779331-48-5
96%
1g
Videos
SDS