missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylbenzoic Acid, 98+%, ACROS Organics™

£11.00 - £128.00

Chemical Identifiers

CAS 85-52-9
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00002472
InChI Key FGTYTUFKXYPTML-UHFFFAOYSA-N
Synonym o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid
PubChem CID 6813
IUPAC Name 2-benzoylbenzoic acid
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10112230 SDS View Documents Acros Organics
105710250
25g Plastic bottle £11.00
25g View alternative products
Save Up to 
»
EU Stock 11 Please to purchase this item. Need a web account? Register with us today!
10070720 SDS View Documents Acros Organics
105715000
500g Plastic bottle £69.50
500g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-03-2020
Please to purchase this item. Need a web account? Register with us today!
10677242 SDS View Documents Acros Organics
105710010
1kg Plastic bottle £128.00
1kg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 03-03-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

85-52-9
226.23
FGTYTUFKXYPTML-UHFFFAOYSA-N
6813
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2C(=O)O
C14H10O3
MFCD00002472
o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid
2-benzoylbenzoic acid

Specifications

2-Benzoylbenzoic acid
Plastic bottle
Beige to White
25g
85-52-9
100.0
C14H10O3
MFCD00002472
o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2C(=O)O
226.23
226.23
98+%
Authentic
257.0°C to 265.0°C
126.0°C to 130.0°C
98+%
98.0
98+%
C6H5COC6H4CO2H
10, 747
FGTYTUFKXYPTML-UHFFFAOYSA-N
2-benzoylbenzoic acid
6813
Crystalline Powder
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Benzoylbenzoic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.