missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoxazolinone, 98%, Thermo Scientific Chemicals

Catalog Number p-3777400
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
59-49-4
C7H5NO2
135.12
MFCD00005716
ASSKVPFEZFQQNQ-UHFFFAOYSA-N
2-benzoxazolinone, benzoxazolinone, benzoxazolone, 2-hydroxybenzoxazole, 2-benzoxazolol, 2 3h-benzoxazolone, 1,3-benzoxazol-2 3h-one, 2-benzoxazolone, benzo d oxazol-2 3h-one, benzoxazolin-2-one
6043
3H-1,3-benzoxazol-2-one
O=C1NC2=CC=CC=C2O1
This item is not returnable. View return policy
59-49-4
C7H5NO2
135.12
MFCD00005716
ASSKVPFEZFQQNQ-UHFFFAOYSA-N
2-benzoxazolinone, benzoxazolinone, benzoxazolone, 2-hydroxybenzoxazole, 2-benzoxazolol, 2 3h-benzoxazolone, 1,3-benzoxazol-2 3h-one, 2-benzoxazolone, benzo d oxazol-2 3h-one, benzoxazolin-2-one
6043
3H-1,3-benzoxazol-2-one
O=C1NC2=CC=CC=C2O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 137°C to 141°C
Color Beige to Brown-Gray
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in ethanol,ethyl ether and,trifluoroacetic acid
Formula Weight 135.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Benzoxazolinone
Show More Show Less
compliance-icons
Product Identifier
  • 2-Benzoxazolinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'