missing translation for 'onlineSavingsMsg'
Learn More

2-Benzothienylboronic acid, 98%, ACROS Organics™

Brand:  Acros Organics 358910250

Code : 29

Additional Details : CAS Number : 98437-23-1 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10733095

  • £178.40 / 25g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

2-Benzothienylboronic acid
97.5
98%
C8H7BO2S
MFCD01075674
98437-23-1
100.0
98%
178.02

Specifications

2-Benzothienylboronic acid
97.5
Powder
98%
178.02
Authentic
White to off-white
25g
98437-23-1
100.0
98%
C8H7BO2S
MFCD01075674
Glass bottle
236.0° to 237.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Benzothienylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS