missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
10 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Absorbance | (c=1, CH2Cl2) (1cm cell), (c=1, CH2Cl2) (1cm cell), 2 max. at 400nm |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 55.0°C to 59.0°C |
Color | White to Beige |
Boiling Point | 102°C to 106°C (0.2 hPa) |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- (#1)-2-Azabicyclo¢2.2.1!hept-5-en-3-one
- Warning
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'