missing translation for 'onlineSavingsMsg'
Learn More

(2-anilinopyrimidin-5-yl)methylamine, 95%, Thermo Scientific™

Catalog Number p-3880540
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1093860-47-9
C11H12N4
200.245
MFCD11841074
IGMKIKUXMJLMOP-UHFFFAOYSA-N
5-aminomethyl-n-phenylpyrimidin-2-amine, 2-anilinopyrimidin-5-yl methylamine, 5-aminomethyl-2-phenylamino pyrimidine, 5-aminomethyl pyrimidin-2-yl phenylamine
43811051
5-(aminomethyl)-N-phenylpyrimidin-2-amine
C1=CC=C(C=C1)NC2=NC=C(C=N2)CN
This item is not returnable. View return policy
1093860-47-9
C11H12N4
200.245
MFCD11841074
IGMKIKUXMJLMOP-UHFFFAOYSA-N
5-aminomethyl-n-phenylpyrimidin-2-amine, 2-anilinopyrimidin-5-yl methylamine, 5-aminomethyl-2-phenylamino pyrimidine, 5-aminomethyl pyrimidin-2-yl phenylamine
43811051
5-(aminomethyl)-N-phenylpyrimidin-2-amine
C1=CC=C(C=C1)NC2=NC=C(C=N2)CN
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 200.24
Percent Purity 95%
Chemical Name or Material (2-Anilinopyrimidin-5-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'