missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-4228346
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
40283-46-3
C4H4N2O2S
144.15
MFCD06203554
ZFMRDDYYJJCBKC-UHFFFAOYSA-N
2-aminothiazole-5-carboxylic acid, 2-amino-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 2-amino, 2-amino-5-carboxy-1,3-thiazole, pubchem21384, acmc-209jcp, 2-aminothiazole-5-carboxylicacid, 5-thiazolecarboxylicacid, 2-amino, 2-aminothiazole-5-formic acid, 2-azanyl-1,3-thiazole-5-carboxylic acid
315243
2-amino-1,3-thiazole-5-carboxylic acid
C1=C(SC(=N1)N)C(=O)O
This item is not returnable. View return policy
40283-46-3
C4H4N2O2S
144.15
MFCD06203554
ZFMRDDYYJJCBKC-UHFFFAOYSA-N
2-aminothiazole-5-carboxylic acid, 2-amino-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 2-amino, 2-amino-5-carboxy-1,3-thiazole, pubchem21384, acmc-209jcp, 2-aminothiazole-5-carboxylicacid, 5-thiazolecarboxylicacid, 2-amino, 2-aminothiazole-5-formic acid, 2-azanyl-1,3-thiazole-5-carboxylic acid
315243
2-amino-1,3-thiazole-5-carboxylic acid
C1=C(SC(=N1)N)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 182.0°C to 184.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 144.15
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminothiazole-5-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminothiazole-5-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'