missing translation for 'onlineSavingsMsg'
Learn More

(2-Aminopyrid-4-yl)methanol, 97%, ACROS Organics™

Brand:  Acros Organics 437370010

Code : 29

Additional Details : CAS Number : 105250-17-7 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 11966371

  • £49.00 / 1g


Estimated Shipment
08-05-2018
Add to basket


Specifications

Chemical Identifiers

(2-Aminopyrid-4-yl)methanol
96.0
97%
C6H8N2O
105250-17-7
100.0
97%
124.14

Specifications

(2-Aminopyrid-4-yl)methanol
Authentic
100.0
White to yellow
Crystalline Powder
97%
C6H8N2O
105250-17-7
96.0
Glass bottle
75.5° to 84.0°C
1g
97%
124.14
Videos
SDS