missing translation for 'onlineSavingsMsg'
Learn More

(2-Aminopyrid-4-yl)methanol, 97%, Thermo Scientific Chemicals

Catalog Number 11976371
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
105250-17-7
C6H9N2O
125.15
MFCD03791261
ZRJJXXDQIQFZBW-UHFFFAOYSA-O
2-aminopyridin-4-yl methanol, 2-amino-pyridin-4-yl-methanol, 2-aminopyridine-4-methanol, 2-amino-4-pyridinylmethanol, 4-pyridinemethanol, 2-amino, 2-aminopyridin-4-yl-methanol, 2-amino-4-pyridinemethanol, 2-amino-4-pyridinyl methanol, 2-amino-4-hydroxymethylpyridine, 2-amino-4-pyridyl methan-1-ol
1515296
(2-aminopyridin-4-yl)methanol
NC1=CC(CO)=CC=[NH+]1
This item is not returnable. View return policy
105250-17-7
C6H9N2O
125.15
MFCD03791261
ZRJJXXDQIQFZBW-UHFFFAOYSA-O
2-aminopyridin-4-yl methanol, 2-amino-pyridin-4-yl-methanol, 2-aminopyridine-4-methanol, 2-amino-4-pyridinylmethanol, 4-pyridinemethanol, 2-amino, 2-aminopyridin-4-yl-methanol, 2-amino-4-pyridinemethanol, 2-amino-4-pyridinyl methanol, 2-amino-4-hydroxymethylpyridine, 2-amino-4-pyridyl methan-1-ol
1515296
(2-aminopyridin-4-yl)methanol
NC1=CC(CO)=CC=[NH+]1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 75.5°C to 84.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 124.14
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (2-Aminopyrid-4-yl)methanol
Show More Show Less
compliance-icons
Product Identifier
  • (2-Aminopyrid-4-yl)methanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'