missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminophenol, 99%, Thermo Scientific Chemicals
17.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.
Solubility
Soluble in water (17 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.
Solubility
Soluble in water (17 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Melting Point | 173°C to 177°C |
Density | 1.328 |
Boiling Point | 150°C (11 mmHg) |
Flash Point | >175°C (347°F) |
Odor | Characteristic |
Quantity | 100 g |
UN Number | UN2512 |
Beilstein | 606075 |
Merck Index | 14,461 |
Solubility Information | Soluble in water (17g/L at 20°C). |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'