missing translation for 'onlineSavingsMsg'
Learn More

2-Aminonicotinic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-57911
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5345-47-1
C6H6N2O2
138.13
MFCD00006318
KPIVDNYJNOPGBE-UHFFFAOYSA-N
2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid
72930
CHEBI:68572
2-aminopyridine-3-carboxylic acid
NC1=NC=CC=C1C(O)=O
This item is not returnable. View return policy
5345-47-1
C6H6N2O2
138.13
MFCD00006318
KPIVDNYJNOPGBE-UHFFFAOYSA-N
2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid
72930
CHEBI:68572
2-aminopyridine-3-carboxylic acid
NC1=NC=CC=C1C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 295.0°C to 297.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula H2N(C5H3N)COOH
Quantity 5 g
Beilstein 22, 542
Solubility Information Solubility in water: soluble.
Formula Weight 138.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminonicotinic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'