missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminomethyl-1H-imidazole dihydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 250.0°C to 251.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5 to max. 102.5% min. (Argentometry) |
Packaging | Glass Bottle |
Quantity | 1 g |
IUPAC Name | 1-(1H-imidazol-2-yl)methanamine dihydrochloride |
Formula Weight | 170.04 |
Percent Purity | 98% |
Chemical Name or Material | 2-Aminomethyl-1H-imidazole dihydrochloride |
Safety and Handling
Product Identifier
- 2-Aminomethyl-1H-imidazole dihydrochloride, 98%
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'