missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)-1-ethylpyrrolidine, 95%, ACROS Organics™

£134.42 - £491.15

Chemical Identifiers

CAS 26116-12-1
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD00003178
InChI Key UNRBEYYLYRXYCG-UHFFFAOYSA-N
Synonym 2-aminomethyl-1-ethylpyrrolidine, 1-ethylpyrrolidin-2-yl methanamine, n-ethyl-2-aminomethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl, 1-ethylpyrrolidin-2-ylmethylamine, 1-ethylpyrrolidin-2-yl methylamine, 1-ethyl-2-aminomethylpyrrolidine, 1-ethyl-2-aminomethyl pyrrolidine, 1-1-ethylpyrrolidin-2-yl methanamine
PubChem CID 117295
IUPAC Name (1-ethylpyrrolidin-2-yl)methanamine
SMILES CCN1CCCC1CN
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10032330
SDS View Documents
Acros Organics
179480100
10g Glass bottle
£134.42
10g
View alternative products
Save Up to 
»
Estimated Shipment
06-05-2021
Please to purchase this item. Need a web account? Register with us today!
10761261
SDS View Documents
Acros Organics
179480500
50g Glass bottle
£491.15
50g
View alternative products
Save Up to 
»
Estimated Shipment
06-05-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

26116-12-1
128.22
UNRBEYYLYRXYCG-UHFFFAOYSA-N
117295
CCN1CCCC1CN
C7H16N2
MFCD00003178
2-aminomethyl-1-ethylpyrrolidine, 1-ethylpyrrolidin-2-yl methanamine, n-ethyl-2-aminomethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl, 1-ethylpyrrolidin-2-ylmethylamine, 1-ethylpyrrolidin-2-yl methylamine, 1-ethyl-2-aminomethylpyrrolidine, 1-ethyl-2-aminomethyl pyrrolidine, 1-1-ethylpyrrolidin-2-yl methanamine
(1-ethylpyrrolidin-2-yl)methanamine

Specifications

2-(Aminomethyl)-1-ethylpyrrolidine
56°C
Glass bottle
0.884
Yellow
95%
94.0
95%
MFCD00003178
UNRBEYYLYRXYCG-UHFFFAOYSA-N
(1-ethylpyrrolidin-2-yl)methanamine
117295
Liquid
0.8840g/mL
Authentic
1.4655 to 1.4675
58.0°C to 60.0°C (16.0mmHg)
10g
26116-12-1
100.0
C7H16N2
2-aminomethyl-1-ethylpyrrolidine, 1-ethylpyrrolidin-2-yl methanamine, n-ethyl-2-aminomethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl, 1-ethylpyrrolidin-2-ylmethylamine, 1-ethylpyrrolidin-2-yl methylamine, 1-ethyl-2-aminomethylpyrrolidine, 1-ethyl-2-aminomethyl pyrrolidine, 1-1-ethylpyrrolidin-2-yl methanamine
CCN1CCCC1CN
128.22
128.22
95%
Safety and Handling

Safety and Handling

Product Identifier
  • 2-(Aminomethyl)-1-ethylpyrrolidine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS