missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl hydrogen Sulfate 99%, ACROS Organics™

Brand:  Acros Organics 103640025

Code : 29

Additional Details : CAS Number : 926-39-6 Weight : 2.55000kg

Packaging Glass bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 10493205

  • £973.00 / 2.50kg


Estimated Shipment
19-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Aminoethyl hydrogen sulfate
926-39-6
100.0
99%
H2NCH2CH2OSO3H
MFCD00008179
99%
98.5
≥98.5%
C2H7NO4S
141.14

Specifications

2-Aminoethyl hydrogen sulfate
926-39-6
100.0
Glass bottle
Fine Crystalline Powder
2.5kg
C2H7NO4S
141.14
04, 276
99%
98.5
Authentic
White to slightly beige
≥98.5%
99%
H2NCH2CH2OSO3H
MFCD00008179
Videos
SDS