missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog Number p-3756757
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5344-90-1
C7H9NO
123.15
MFCD00007749
VYFOAVADNIHPTR-UHFFFAOYSA-N
2-aminobenzyl alcohol, 2-aminophenyl methanol, 2-aminobenzylalcohol, o-aminobenzyl alcohol, 2-aminobenzenemethanol, benzenemethanol, 2-amino, o-aminobenzylic alcohol, o-hydroxymethyl aniline, benzyl alcohol, o-amino, 2-hydroxymethylaniline
21439
(2-aminophenyl)methanol
C1=CC=C(C(=C1)CO)N
This item is not returnable. View return policy
5344-90-1
C7H9NO
123.15
MFCD00007749
VYFOAVADNIHPTR-UHFFFAOYSA-N
2-aminobenzyl alcohol, 2-aminophenyl methanol, 2-aminobenzylalcohol, o-aminobenzyl alcohol, 2-aminobenzenemethanol, benzenemethanol, 2-amino, o-aminobenzylic alcohol, o-hydroxymethyl aniline, benzyl alcohol, o-amino, 2-hydroxymethylaniline
21439
(2-aminophenyl)methanol
C1=CC=C(C(=C1)CO)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 82.0°C to 86.0°C
Color Beige to Brown-Gray
Boiling Point 162.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CH2OH
Refractive Index 1.4905
Quantity 10 g
Beilstein 13, 615
Solubility Information Solubility in water: 30g/100g (20°C). Other solubilities: 40g/100g thf (14°C),0.64g/100g toluene (20°C),soluble in chlorinated solvents,soluble in ethanol,ether,very soluble in benzene
Formula Weight 123.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'