missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminobenzyl alcohol, 98%, Thermo Scientific Chemicals
12.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
250 g
Description
2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines. It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2 to yield 2-aminobenzaldehyde. was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines. It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2 to yield 2-aminobenzaldehyde. was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.
Solubility
Very soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines. It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2 to yield 2-aminobenzaldehyde. was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.
Solubility
Very soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Melting Point | 81°C to 84°C |
Boiling Point | 160°C (10 mmHg) |
Quantity | 10 g |
Beilstein | 1072211 |
Sensitivity | Air and light sensitive |
Solubility Information | Very soluble in water. |
Formula Weight | 123.16 |
Percent Purity | 98% |
Chemical Name or Material | 2-Aminobenzyl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'