missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzothiazole, 97%, Thermo Scientific Chemicals

Catalog Number 10503891
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
136-95-8
C7H6N2S
150.2
MFCD00005785
UHGULLIUJBCTEF-UHFFFAOYSA-N
2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
8706
1,3-benzothiazol-2-amine
C1=CC=C2C(=C1)N=C(S2)N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
136-95-8
C7H6N2S
150.2
MFCD00005785
UHGULLIUJBCTEF-UHFFFAOYSA-N
2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
8706
1,3-benzothiazol-2-amine
C1=CC=C2C(=C1)N=C(S2)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 129°C to 134°C
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Quantity 500 g
Beilstein 27, 182
Merck Index 15, 419
Solubility Information Solubility in water: sparingly soluble in water. Other solubilities: soluble in concentrated acids,freely soluble in alcohol,ether and chloroform
Formula Weight 150.2
Percent Purity 97%
Physical Form Powder or Flakes
Chemical Name or Material 2-Aminobenzothiazole, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'