missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzothiazole-6-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7030686
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
93-85-6
C8H6N2O2S
194.21
MFCD00054180
ZEAKWWWXCZMODH-UHFFFAOYSA-N
2-aminobenzothiazole-6-carboxylic acid, 2-aminobenzo d thiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, 2-amino, 2-amino-benzothiazole-6-carboxylic acid, 2-amino-6-benzothiazolecarboxylic acid, asischem b67429, zlchem 188, akos bb-7723, salor-int l481106-1ea, ksc487a1l
66740
2-amino-1,3-benzothiazole-6-carboxylic acid
NC1=NC2=CC=C(C=C2S1)C(O)=O
This item is not returnable. View return policy
93-85-6
C8H6N2O2S
194.21
MFCD00054180
ZEAKWWWXCZMODH-UHFFFAOYSA-N
2-aminobenzothiazole-6-carboxylic acid, 2-aminobenzo d thiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, 2-amino, 2-amino-benzothiazole-6-carboxylic acid, 2-amino-6-benzothiazolecarboxylic acid, asischem b67429, zlchem 188, akos bb-7723, salor-int l481106-1ea, ksc487a1l
66740
2-amino-1,3-benzothiazole-6-carboxylic acid
NC1=NC2=CC=C(C=C2S1)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Formula Weight 194.21
Percent Purity 95%
Quantity 1 g
Chemical Name or Material 2-Aminobenzothiazole-6-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'