missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxybenzothiazole, 98%, ACROS Organics™

Brand:  Acros Organics 172022500

Code : 29

Additional Details : CAS Number : 1747-60-0 Weight : 0.30000kg


Quantity 250g
Packaging Glass bottle

 View more versions of this product

Product Code. 10791251

  • £209.60 / 250g


Estimated Shipment
22-06-2018
Add to basket


Specifications

Chemical Identifiers

2-Amino-6-methoxybenzothiazole
1747-60-0
100.0
98%
180.23
98%
97.5
≥97.5%
C8H8N2OS
MFCD00005787

Specifications

2-Amino-6-methoxybenzothiazole
1747-60-0
100.0
Glass bottle
≥97.5%
C8H8N2OS
180.23
White to beige or grayish-lilac
27, II, 334
98%
97.5
Authentic
Crystalline Powder
98%
163° to 168°C
MFCD00005787
250g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Amino-6-methoxybenzothiazole

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS