missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-hydroxy-8-mercaptopurine, 97%, Thermo Scientific™

Catalog Number p-7044686
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
28128-40-7
C5H5N5OS
183.189
MFCD00075623
JHEKNTQSGTVPAO-UHFFFAOYSA-N
2-amino-6-hydroxy-8-mercaptopurine, 8-mercaptoguanine, 2-amino-8-mercapto-7h-purin-6-ol, 2-amino-8-sulfanyl-1,9-dihydro-6h-purin-6-one, 2-amino-1,7,8,9-tetrahydro-2-thioxo-6h-purin-6-one, 2-amino-8-mercapto-1,9-dihydro-purin-6-one, 2-amino-8-thioxo-3,7-dihydropurin-6-one, 2-amino-8-sulfanyl-7h-purin-6-ol, 2-amino-8-sulfanyl-9h-purin-6-ol, 2-amino-8-thioxo-1,7,8,9-tetrahydro-purin-6-one
2725005
2-amino-8-sulfanylidene-7,9-dihydro-3H-purin-6-one
C12=C(NC(=NC1=O)N)NC(=S)N2
This item is not returnable. View return policy
28128-40-7
C5H5N5OS
183.189
MFCD00075623
JHEKNTQSGTVPAO-UHFFFAOYSA-N
2-amino-6-hydroxy-8-mercaptopurine, 8-mercaptoguanine, 2-amino-8-mercapto-7h-purin-6-ol, 2-amino-8-sulfanyl-1,9-dihydro-6h-purin-6-one, 2-amino-1,7,8,9-tetrahydro-2-thioxo-6h-purin-6-one, 2-amino-8-mercapto-1,9-dihydro-purin-6-one, 2-amino-8-thioxo-3,7-dihydropurin-6-one, 2-amino-8-sulfanyl-7h-purin-6-ol, 2-amino-8-sulfanyl-9h-purin-6-ol, 2-amino-8-thioxo-1,7,8,9-tetrahydro-purin-6-one
2725005
2-amino-8-sulfanylidene-7,9-dihydro-3H-purin-6-one
C12=C(NC(=NC1=O)N)NC(=S)N2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 1 g
Beilstein 982327
Formula Weight 183.19
Percent Purity 97%
Chemical Name or Material 2-Amino-6-hydroxy-8-mercaptopurine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'