missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-chlorobenzothiazole, 99%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 197°C to 201°C |
Color | White to Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 10 g |
Formula Weight | 184.65 |
Percent Purity | 99% |
Show More |
Safety and Handling
Product Identifier
- 2-Amino-6-chlorobenzothiazole
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'