missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloro-4-pyrimidinol Monohydrate 95%, ACROS Organics™

Brand:  Acros Organics 103430050

Code : 29

Additional Details : CAS Number : 206658-81-3 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10477470

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2-Amino-6-chloro-4-pyrimidinol hydrate
206658-81-3
100.0
95%
145.55
95%
94.0
≥94% (HPLC)
C4H4ClN3O·xH2O
MFCD00149407

Specifications

2-Amino-6-chloro-4-pyrimidinol hydrate
Glass bottle
Authentic
100.0
5g
≥94% (HPLC)
C4H4ClN3O·xH2O
MFCD00149407
95%
206658-81-3
94.0
Light yellow
Powder
95%
145.55
Videos
SDS