missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-6-chloro-4-(dimethylamino)pyrimidine, 97%

Brand:  Alfa Aesar™ H64847.06

Code : NEW

Additional Details : CAS Number : 1007-11-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15472239

  • £298.92 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Amino-6-chloro-4-(dimethylamino)pyrimidine
172.62
6221
1007-11-0
MFCD13188630
6-Chloro-N4,N4-dimethylpyrimidine-2,4-diamine

Specifications

2-Amino-6-chloro-4-(dimethylamino)pyrimidine
162° to 165°C
Solid
172.62
6221
1007-11-0
5g
97%
MFCD13188630
6-Chloro-N4,N4-dimethylpyrimidine-2,4-diamine
Videos
SDS