missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-6-bromobenzonitrile, 95%

Brand:  Alfa Aesar™ H61927.03

Code : NEW

Additional Details : CAS Number : 77326-62-6 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15438518

  • £35.50 / 1g


Estimated Shipment date
from Supplier 23-03-2018
Add to basket

Specifications

Chemical Identifiers

2-Amino-6-bromobenzonitrile
197.02
UN3439
77326-62-6
MFCD09834769

Specifications

2-Amino-6-bromobenzonitrile
Powder
Off-white
1g
UN3439
77326-62-6
95%
197.02
MFCD09834769
Videos
SDS