missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-bromo-3-nitropyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 203.0°C to 204.0°C |
Assay Percent Range | 96% min. (HPLC) |
Quantity | 5 g |
Synonym | 2-amino-6-bromo-3-nitropyridine, 6-bromo-3-nitro-2-pyridinamine, 2-pyridinamine, 6-bromo-3-nitro, 6-bromo-3-nitro-pyridin-2-ylamine, 6-bromo-3-nitro-2-pyridylamine, 2-pyridinamine,6-bromo-3-nitro, acmc-209puv, chempacific 38154, 6-amino-2-bromo-5-nitropyridine, 2-amino-3-nitro-6-bromopyridine |
Formula Weight | 218.01 |
Percent Purity | 97% |
Chemical Name or Material | 2-Amino-6-bromo-3-nitropyridine |
Safety and Handling
Product Identifier
- 2-Amino-6-bromo-3-nitropyridine
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'