missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-thien-2-ylbenzoic acid, 97%, Thermo Scientific™

Catalog Number p-4228345
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
885268-32-6
C11H9NO2S
219.26
PXSSPVWKCSZVKQ-UHFFFAOYSA-N
16768750
2-amino-5-thiophen-2-ylbenzoic acid
C1=CSC(=C1)C2=CC(=C(C=C2)N)C(=O)O
This item is not returnable. View return policy
885268-32-6
C11H9NO2S
219.26
PXSSPVWKCSZVKQ-UHFFFAOYSA-N
16768750
2-amino-5-thiophen-2-ylbenzoic acid
C1=CSC(=C1)C2=CC(=C(C=C2)N)C(=O)O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 209.4°C to 210°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Synonym 2-amino-5-thiophen-2-yl benzoic acid, 2-amino-5-thien-2-ylbenzoic acid, 2-amino-5-2-thienyl benzoic acid, benzoicacid, 2-amino-5-2-thienyl, benzoicacid,2-amino-5-2-thienyl
Formula Weight 219.26
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-thien-2-ylbenzoic acid
Show More Show Less
Product Identifier
  • 2-Amino-5-thien-2-ylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'