missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-thien-2-ylbenzoic acid, 97%, ACROS Organics™

Brand:  Acros Organics 432770050

Code : 29

Additional Details : CAS Number : 885268-32-6 Weight : 0.05500kg

Quantity 5g

 View more versions of this product

Product Code. 10471440

  • £274.20 / 5g


Estimated Shipment
15-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Amino-5-thien-2-ylbenzoic acid
96.0
≥96.0%
C11H9NO2S
885268-32-6
100.0
97%
219.26

Specifications

2-Amino-5-thien-2-ylbenzoic acid
96.0
Authentic
≥96.0%
C11H9NO2S
219.26
885268-32-6
100.0
Crystalline Powder
97%
Yellow
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Amino-5-thien-2-ylbenzoic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS