missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzotrifluoride, 98%, Thermo Scientific™

Catalog Number p-57835
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
121-01-7
MFCD00007365
HOTZLWVITTVZGY-UHFFFAOYSA-N
2-amino-5-nitrobenzotrifluoride, 4-nitro-2-trifluoromethyl aniline, 2-amino-5-nitro-trifluorobenzene, 4-nitro-alpha,alpha,alpha-trifluoro-o-toluidine, 4-nitro-2-trifluoromethyl benzenamine, 4-nitro-a,a,a-trifluoro-o-toluidine, benzenamine, 4-nitro-2-trifluoromethyl, 5-nitro-2-aminobenzotrifluoride, 2-trifluoromethyl-4-nitroaniline, a,a,a-trifluoro-4-nitro-o-toluidine
67128
4-nitro-2-(trifluoromethyl)aniline
C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)N
This item is not returnable. View return policy
121-01-7
MFCD00007365
HOTZLWVITTVZGY-UHFFFAOYSA-N
2-amino-5-nitrobenzotrifluoride, 4-nitro-2-trifluoromethyl aniline, 2-amino-5-nitro-trifluorobenzene, 4-nitro-alpha,alpha,alpha-trifluoro-o-toluidine, 4-nitro-2-trifluoromethyl benzenamine, 4-nitro-a,a,a-trifluoro-o-toluidine, benzenamine, 4-nitro-2-trifluoromethyl, 5-nitro-2-aminobenzotrifluoride, 2-trifluoromethyl-4-nitroaniline, a,a,a-trifluoro-4-nitro-o-toluidine
67128
4-nitro-2-(trifluoromethyl)aniline
C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)N
CAS Min % 97.5
CAS Max % 100.0
Melting Point 90°C to 93°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H5F3N2O2
Linear Formula H2NC6H3(NO2)CF3
Quantity 5 g
Molecular Weight (g/mol) 206.12
Formula Weight 206.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-5-nitrobenzotrifluoride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-nitrobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'