missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-mercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals

Catalog Number p-7081460
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2349-67-9
C2H3N3S2
133.187
MFCD00003108
GDGIVSREGUOIJZ-UHFFFAOYSA-N
5-amino-1,3,4-thiadiazole-2-thiol, 2-amino-5-mercapto-1,3,4-thiadiazole, usaf pd-25, 1,3,4-thiadiazole-2 3h-thione, 5-amino, unii-b1heg7v21s, 5-amino-1,3,4-thiadiazole-2 3h-thione, 2-amino-1,3,4-thiadiazole-5-thiol, 2-thiol-5-amino-1,3,4-thiadiazole, ccris 6893, 2-mercapto-5-amino-1,3,4-thiadiazole
2723847
5-amino-3H-1,3,4-thiadiazole-2-thione
C1(=NNC(=S)S1)N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2349-67-9
C2H3N3S2
133.187
MFCD00003108
GDGIVSREGUOIJZ-UHFFFAOYSA-N
5-amino-1,3,4-thiadiazole-2-thiol, 2-amino-5-mercapto-1,3,4-thiadiazole, usaf pd-25, 1,3,4-thiadiazole-2 3h-thione, 5-amino, unii-b1heg7v21s, 5-amino-1,3,4-thiadiazole-2 3h-thione, 2-amino-1,3,4-thiadiazole-5-thiol, 2-thiol-5-amino-1,3,4-thiadiazole, ccris 6893, 2-mercapto-5-amino-1,3,4-thiadiazole
2723847
5-amino-3H-1,3,4-thiadiazole-2-thione
C1(=NNC(=S)S1)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼243°C (decomposition)
Quantity 25 g
Beilstein 81728
Formula Weight 133.19
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-mercapto-1,3,4-thiadiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'