missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-iodobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3777279
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5326-47-6
C7H6INO2
263.03
MFCD00007849
GOLGILSVWFKZRQ-UHFFFAOYSA-N
5-iodoanthranilic acid, benzoic acid, 2-amino-5-iodo, 5-iodoanthranil acid, anthranilic acid, 5-iodo, 2-amino-5-iodo-benzoic acid, pubchem2543, 5-iodo anthranilic acid, acmc-1ays8, 2-amino-5-iodobenzoicacid, 5-iodo-2-aminobenzoic acid
72911
2-amino-5-iodobenzoic acid
C1=CC(=C(C=C1I)C(=O)O)N
This item is not returnable. View return policy
5326-47-6
C7H6INO2
263.03
MFCD00007849
GOLGILSVWFKZRQ-UHFFFAOYSA-N
5-iodoanthranilic acid, benzoic acid, 2-amino-5-iodo, 5-iodoanthranil acid, anthranilic acid, 5-iodo, 2-amino-5-iodo-benzoic acid, pubchem2543, 5-iodo anthranilic acid, acmc-1ays8, 2-amino-5-iodobenzoicacid, 5-iodo-2-aminobenzoic acid
72911
2-amino-5-iodobenzoic acid
C1=CC(=C(C=C1I)C(=O)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 219.0°C to 221.0°C
Color Beige or Gray to Purple
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NCH3(I)CO2H
Quantity 25 g
Beilstein 14, 373
Formula Weight 263.03
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-iodobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-iodobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'