missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-hydroxypyrimidine, 95%, Thermo Scientific Chemicals

Catalog Number p-4229060
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
143489-45-6
C4H5N3O
111.10
MFCD08062411
NVYMOVCPYONOSF-UHFFFAOYSA-N
2-amino-5-hydroxypyrimidine, 5-pyrimidinol, 2-amino, 2-amino-pyrimidin-5-ol, 5-hydroxypyrimidin-2-amine, 2-amino-5-hydroxy pyrimidine, 2-amino-5-pyrimidinol, zlchem 1241, pubchem21503, 5-hydroxy-2-aminopyrimidine, acmc-1c7b8
17750388
2-aminopyrimidin-5-ol
NC1=NC=C(O)C=N1
This item is not returnable. View return policy
143489-45-6
C4H5N3O
111.10
MFCD08062411
NVYMOVCPYONOSF-UHFFFAOYSA-N
2-amino-5-hydroxypyrimidine, 5-pyrimidinol, 2-amino, 2-amino-pyrimidin-5-ol, 5-hydroxypyrimidin-2-amine, 2-amino-5-hydroxy pyrimidine, 2-amino-5-pyrimidinol, zlchem 1241, pubchem21503, 5-hydroxy-2-aminopyrimidine, acmc-1c7b8
17750388
2-aminopyrimidin-5-ol
NC1=NC=C(O)C=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 198.6°C to 199°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 111.1
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Amino-5-hydroxypyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-hydroxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'