missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluorobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-3756799
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
446-08-2
C7H6FNO2
155.13
MFCD00055566
FPQMGQZTBWIHDN-UHFFFAOYSA-N
5-fluoroanthranilic acid, 2-amino-5-fluoro-benzoic acid, benzoic acid, 2-amino-5-fluoro, 5-faa, 5-fluoro-2-aminobenzoic acid, 4own, 5rg, pubchem1322, 5-fluoroanthanilic acid, acmc-209jyo
101412
CHEBI:78042
2-amino-5-fluorobenzoic acid
C1=CC(=C(C=C1F)C(=O)O)N
This item is not returnable. View return policy
446-08-2
C7H6FNO2
155.13
MFCD00055566
FPQMGQZTBWIHDN-UHFFFAOYSA-N
5-fluoroanthranilic acid, 2-amino-5-fluoro-benzoic acid, benzoic acid, 2-amino-5-fluoro, 5-faa, 5-fluoro-2-aminobenzoic acid, 4own, 5rg, pubchem1322, 5-fluoroanthanilic acid, acmc-209jyo
101412
CHEBI:78042
2-amino-5-fluorobenzoic acid
C1=CC(=C(C=C1F)C(=O)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 181°C to 185°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(F)COOH
Quantity 1 g
Formula Weight 155.13
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Amino-5-fluorobenzoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'