missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluoro-4-methylpyridine, 95%, Thermo Scientific Chemicals

Catalog Number p-7041704
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
301222-66-2
C6H7FN2
126.134
MFCD06659479
RKMOPXJNMQWPLH-UHFFFAOYSA-N
2-amino-5-fluoro-4-picoline, 2-amino-5-fluoro-4-methylpyridine, 5-fluoro-4-methyl-2-pyridinamine, 2-amino-4-methyl-5-fluoropyridine, pubchem1256, 5-fluoro-4-methyl-pyridin-2-amine, 2-pyridinamine,5-fluoro-4-methyl, 2-amino-4-methyl-5-fluoro pyridine, e 5-fluoro-4-methyl-2-pyridinamine, 2-pyridinamine, 5-fluoro-4-methyl-9ci
22063670
5-fluoro-4-methylpyridin-2-amine
CC1=CC(=NC=C1F)N
This item is not returnable. View return policy
301222-66-2
C6H7FN2
126.134
MFCD06659479
RKMOPXJNMQWPLH-UHFFFAOYSA-N
2-amino-5-fluoro-4-picoline, 2-amino-5-fluoro-4-methylpyridine, 5-fluoro-4-methyl-2-pyridinamine, 2-amino-4-methyl-5-fluoropyridine, pubchem1256, 5-fluoro-4-methyl-pyridin-2-amine, 2-pyridinamine,5-fluoro-4-methyl, 2-amino-4-methyl-5-fluoro pyridine, e 5-fluoro-4-methyl-2-pyridinamine, 2-pyridinamine, 5-fluoro-4-methyl-9ci
22063670
5-fluoro-4-methylpyridin-2-amine
CC1=CC(=NC=C1F)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 9694924
Formula Weight 126.13
Percent Purity 95%
Chemical Name or Material 2-Amino-5-fluoro-4-methylpyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'