missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-cyanopyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-2689903
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4214-73-7
C6H6N3
120.13
MFCD00128902
KDVBYUUGYXUXNL-UHFFFAOYSA-O
2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
818260
6-aminopyridine-3-carbonitrile
NC1=CC=C(C=[NH+]1)C#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4214-73-7
C6H6N3
120.13
MFCD00128902
KDVBYUUGYXUXNL-UHFFFAOYSA-O
2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
818260
6-aminopyridine-3-carbonitrile
NC1=CC=C(C=[NH+]1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 159°C to 165°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 119.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-cyanopyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-cyanopyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'