missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-chlorothiazole hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
2-Amino-5-chlorothiazole hydrochloride is used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole. It is used as reactant for preparation of biologically active thiazole derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-5-chlorothiazole hydrochloride is used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole. It is used as reactant for preparation of biologically active thiazole derivatives.
Solubility
Soluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
2-Amino-5-chlorothiazole hydrochloride is used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole. It is used as reactant for preparation of biologically active thiazole derivatives.
Solubility
Soluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications
Melting Point | 168°C to 172°C |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 171.05 |
Percent Purity | 97% |
Chemical Name or Material | 2-Amino-5-chlorothiazole hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'