missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number 11996031
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1073371-77-3
C12H17BClNO2
253.53
MFCD06795672
GVJZHGCVSYBPIM-UHFFFAOYSA-N
4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid, pinacol ester, 2-amino-5-chlorophenylboronic acid pinacol ester, 4-chloro-2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid,pinacol ester, 2-amino-5-chlorobenzeneboronic acid, pinacol ester, 4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine
17750205
4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
CC1(C)OB(OC1(C)C)C1=C(N)C=CC(Cl)=C1
This item is not returnable. View return policy
1073371-77-3
C12H17BClNO2
253.53
MFCD06795672
GVJZHGCVSYBPIM-UHFFFAOYSA-N
4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid, pinacol ester, 2-amino-5-chlorophenylboronic acid pinacol ester, 4-chloro-2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid,pinacol ester, 2-amino-5-chlorobenzeneboronic acid, pinacol ester, 4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine
17750205
4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
CC1(C)OB(OC1(C)C)C1=C(N)C=CC(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 91°C to 93°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 253.54
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chlorophenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 2-Amino-5-chlorophenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'