missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorophenol, 98%, Thermo Scientific Chemicals

Catalog Number p-3756775
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
28443-50-7
C6H6ClNO
143.57
MFCD02093863
FZCQMIRJCGWWCL-UHFFFAOYSA-N
4-chloro-2-hydroxyaniline, phenol, 2-amino-5-chloro, 2-amino-5-chloro-phenol, 5-chloro-2-aminophenol, pubchem1478, 5-chloro-o-aminophenol, 5-chloro-2-amino phenol, 2-hydroxy-4-chloroaniline, acmc-209h2j, 2-azanyl-5-chloranyl-phenol
91591
CHEBI:75051
2-amino-5-chlorophenol
NC1=CC=C(Cl)C=C1O
This item is not returnable. View return policy
28443-50-7
C6H6ClNO
143.57
MFCD02093863
FZCQMIRJCGWWCL-UHFFFAOYSA-N
4-chloro-2-hydroxyaniline, phenol, 2-amino-5-chloro, 2-amino-5-chloro-phenol, 5-chloro-2-aminophenol, pubchem1478, 5-chloro-o-aminophenol, 5-chloro-2-amino phenol, 2-hydroxy-4-chloroaniline, acmc-209h2j, 2-azanyl-5-chloranyl-phenol
91591
CHEBI:75051
2-amino-5-chlorophenol
NC1=CC=C(Cl)C=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 152°C to 157°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in sol. of acids and bases,soluble in ethanol and chloroform
Formula Weight 143.57
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Amino-5-chlorophenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-chlorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'